Assay Details
Target Gene Details
Entrez Gene ID: | 2861 |
Gene Name: | G protein-coupled receptor 37 |
Gene Aliases: |
EDNRBL, PAELR, hET(B)R-LP |
Location: |
Chr.7:124745601-124766025 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR37 | NM_005302.3 | 2 | 2466 | NP_005293.1 |
| AF017262.1 | 2 | 1434 | AAB70008.1 | |
| AF502281.2 | 2 | 1417 | AAM18625.2 | |
| BC040007.1 | 2 | 2068 | AAH40007.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1027091 | Chr.7:124671688 - 124748854 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map