Assay Details
Target Gene Details
Entrez Gene ID: | 285613 |
Gene Name: | RELT like 2 |
Gene Aliases: |
C5orf16 |
Location: |
Chr.5:141636946-141641077 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RELL2 | NM_001130029.1 | NP_001123501.1 | ||
| NM_173828.4 | 1 | 1117 | NP_776189.3 | |
| XM_005268414.3 | XP_005268471.1 | |||
| XM_011537624.1 | XP_011535926.1 | |||
| XM_011537625.2 | 1 | 1121 | XP_011535927.1 | |
| AK054889.1 | 1 | 1117 | ||
| AK075064.1 | BAC11380.1 | |||
| AY358163.1 | 1 | 1053 | AAQ88530.1 | |
| BC063469.1 | AAH63469.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1154058 | Chr.5:141632251 - 142199943 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map