Assay Details
Target Gene Details
Entrez Gene ID: | 23158 |
Gene Name: | TBC1 domain family member 9 |
Gene Aliases: |
GRAMD9, MDR1 |
Location: |
Chr.4:140620782-140756317 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D9 | NM_015130.2 | NP_055945.2 | ||
| AB020689.2 | ||||
| AF131785.1 | ||||
| AK127020.1 | BAC86789.1 | |||
| BC146758.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830089 | Chr.4:140603540 - 140827809 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map