Assay Details
Target Gene Details
Entrez Gene ID: | 84101 |
Gene Name: | ubiquitin specific peptidase 44 |
Gene Aliases: |
- |
Location: |
Chr.12:95516560-95551529 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP44 | NM_001042403.2 | 6 | 2775 | NP_001035862.1 |
| NM_001278393.1 | 6 | 2991 | NP_001265322.1 | |
| NM_032147.4 | 6 | 2920 | NP_115523.2 | |
| NR_125360.1 | 4 | 2505 | ||
| XM_005269172.1 | 7 | 3007 | XP_005269229.1 | |
| XM_005269173.1 | 7 | 3084 | XP_005269230.1 | |
| XM_005269174.1 | 7 | 2851 | XP_005269231.1 | |
| XM_011538800.2 | 8 | 3034 | XP_011537102.1 | |
| XM_011538801.1 | 8 | 2894 | XP_011537103.1 | |
| XM_011538802.1 | 7 | 2854 | XP_011537104.1 | |
| XM_011538803.1 | 7 | 2859 | XP_011537105.1 | |
| XM_011538804.2 | 7 | 3067 | XP_011537106.1 | |
| XM_011538805.1 | 7 | 3708 | XP_011537107.1 | |
| XM_017020009.1 | 6 | 2909 | XP_016875498.1 | |
| XM_017020010.1 | 6 | 2761 | XP_016875499.1 | |
| XM_017020011.1 | 7 | 2787 | XP_016875500.1 | |
| AK027434.1 | 6 | 2725 |
More Information
Set Membership: |
|

Genomic Map