Assay Details
Target Gene Details
Entrez Gene ID: | 7105 |
Gene Name: | tetraspanin 6 |
Gene Aliases: |
T245, TM4SF6, TSPAN-6 |
Location: |
Chr.X:100627108-100637104 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSPAN6 | NM_001278740.1 | 2 | 676 | NP_001265669.1 |
| NM_001278741.1 | 2 | 446 | NP_001265670.1 | |
| NM_001278742.1 | 2 | 446 | NP_001265671.1 | |
| NM_001278743.1 | 2 | 446 | NP_001265672.1 | |
| NM_003270.3 | 2 | 384 | NP_003261.1 | |
| XM_011531018.1 | 3 | 186 | XP_011529320.1 | |
| AB097016.1 | 2 | 280 | BAC77369.1 | |
| AB209876.1 | 2 | 343 | BAD93113.1 | |
| AF043906.1 | 2 | 288 | AAC64257.1 | |
| AF053453.1 | 2 | 240 | AAC69710.1 | |
| AF133426.1 | 2 | 247 | AAF08365.1 | |
| AK075072.1 | 2 | 333 | ||
| AK299176.1 | ||||
| AK300852.1 | ||||
| AK309581.1 | 2 | 659 | ||
| AK315773.1 | 2 | 365 | ||
| AY358825.1 | 2 | 261 | AAQ89184.1 | |
| BC012389.1 | 2 | 324 | AAH12389.1 | |
| BI458635.1 | 3 | 498 | ||
| BT006977.1 | 2 | 221 | AAP35623.1 | |
| CN274931.1 | 2 | 166 | ||
| CR457115.1 | 2 | 221 | CAG33396.1 | |
| CX760536.1 | 2 | 384 | ||
| CX868833.1 | 2 | 155 | ||
| DA038474.1 | 2 | 183 | ||
| U84895.1 | 2 | 288 | AAD00560.1 |
More Information
Set Membership: |
|

Genomic Map