Assay Details
Target Gene Details
Entrez Gene ID: | 2266 |
Gene Name: | fibrinogen gamma chain |
Gene Aliases: |
- |
Location: |
Chr.4:154604134-154612808 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGG | NM_000509.5 | NP_000500.2 | ||
| AA936960.1 | ||||
| AK290824.1 | ||||
| AW014527.1 | ||||
| BC007044.1 | AAH07044.1 | |||
| BC021674.1 | AAH21674.1 | |||
| BX538098.1 | CAD98014.1 |
More Information
Set Membership: |
|

Genomic Map