Assay Details
Target Gene Details
Entrez Gene ID: | 5019 |
Gene Name: | 3-oxoacid CoA-transferase 1 |
Gene Aliases: |
OXCT, SCOT |
Location: |
Chr.5:41730065-41870689 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OXCT1 | NM_000436.3 | 17 | 3141 | NP_000427.1 |
| XM_017009494.1 | 16 | 2932 | XP_016864983.1 | |
| AK125115.1 | 1 | 3718 | ||
| BC009001.1 | 17 | 2865 | AAH09001.1 | |
| U62961.1 | 17 | 2908 | AAB07366.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3604851 | Chr.5:41591779 - 41992753 on Build GRCh38 | Gain |
|
| nsv830271 | Chr.5:41716425 - 41895608 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map