Assay Details
Target Gene Details
Entrez Gene ID: | 5571 |
Gene Name: | protein kinase AMP-activated non-catalytic subunit gamma 1 |
Gene Aliases: |
AMPKG |
Location: |
Chr.12:49002272-49018975 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKAG1 | NM_001206709.1 | NP_001193638.1 | ||
| NM_001206710.1 | NP_001193639.1 | |||
| NM_002733.4 | NP_002724.1 | |||
| XM_005269019.4 | XP_005269076.1 | |||
| XM_005269020.2 | XP_005269077.1 | |||
| XM_006719499.2 | XP_006719562.1 | |||
| XM_011538562.2 | XP_011536864.1 | |||
| XM_017019622.1 | XP_016875111.1 | |||
| XM_017019623.1 | XP_016875112.1 | |||
| AK097606.1 | BAC05117.1 | |||
| AK293332.1 | ||||
| AK303276.1 | ||||
| AK307994.1 | ||||
| AK310250.1 | ||||
| BC000358.2 | AAH00358.1 | |||
| BG772337.1 | ||||
| BG772412.1 | ||||
| BT007345.1 | AAP36009.1 | |||
| CB124830.1 | ||||
| DB020277.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832404 | Chr.12:48864777 - 49064772 on Build GRCh38 | Gain |
|
| nsv558833 | Chr.12:48965097 - 49085029 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map