Assay Details
Target Gene Details
Entrez Gene ID: | 1593 |
Gene Name: | cytochrome P450 family 27 subfamily A member 1 |
Gene Aliases: |
CP27, CTX, CYP27 |
Location: |
Chr.2:218781006-218815293 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP27A1 | NM_000784.3 | 1 | 614 | NP_000775.1 |
| XM_017003488.1 | 1 | 1357 | XP_016858977.1 | |
| AK226039.1 | 1 | 614 | ||
| AK290418.1 | 1 | 220 | ||
| AK310782.1 | 1 | 187 | ||
| AY178622.1 | 1 | 180 | AAO21126.1 | |
| BC040430.1 | 1 | 194 | AAH40430.1 | |
| BC051851.1 | 1 | 192 | AAH51851.1 | |
| M62401.1 | 1 | 201 | AAA52142.1 | |
| X59812.1 | 1 | 381 | CAA42481.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518750 | Chr.2:218751245 - 218804652 on Build GRCh38 | Loss |
|
| esv3594349 | Chr.2:218756892 - 218789452 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map