Assay Details
Target Gene Details
Entrez Gene ID: | 9021 |
Gene Name: | suppressor of cytokine signaling 3 |
Gene Aliases: |
ATOD4, CIS3, Cish3, SOCS-3, SSI-3, SSI3 |
Location: |
Chr.17:78356777-78360079 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOCS3 | NM_003955.4 | 1 | 280 | NP_003946.3 |
| AK309756.1 | 1 | 278 | ||
| AK314212.1 | 1 | 278 | ||
| AU134017.1 | 1 | 278 | ||
| BC060858.1 | 1 | 278 | AAH60858.1 | |
| BE245649.1 | 1 | 98 | ||
| DA671124.1 | 1 | 280 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1112715 | Chr.17:78359819 - 78360519 on Build GRCh38 | Deletion |
|
| nsv833555 | Chr.17:78356467 - 78461269 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map