Assay Details
Target Gene Details
Entrez Gene ID: | 1146 |
Gene Name: | cholinergic receptor nicotinic gamma subunit |
Gene Aliases: |
ACHRG |
Location: |
Chr.2:232539727-232546328 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRNG | NM_005199.4 | 7 | 715 | NP_005190.4 |
| AK125362.1 | 7 | 715 | ||
| BC111802.1 | 6 | 538 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv821918 | Chr.2:232319063 - 232560965 on Build GRCh38 | Gain |
|
| nsv3212 | Chr.2:232526220 - 232564112 on Build GRCh38 | Deletion |
|
| nsv953202 | Chr.2:232474691 - 232546890 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map