Assay Details
Target Gene Details
Entrez Gene ID: | 2528 |
Gene Name: | fucosyltransferase 6 |
Gene Aliases: |
FCT3A, FT1A, Fuc-TVI, FucT-VI |
Location: |
Chr.19:5830626-5839753 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FUT6 | NM_000150.2 | NP_000141.1 | ||
| NM_001040701.1 | NP_001035791.1 | |||
| XM_005259526.4 | XP_005259583.1 | |||
| XM_005259527.3 | XP_005259584.1 | |||
| XM_011527868.2 | XP_011526170.1 | |||
| XM_011527869.2 | XP_011526171.1 | |||
| XM_011527870.2 | XP_011526172.1 | |||
| XM_011527872.2 | XP_011526174.1 | |||
| XM_011527873.2 | XP_011526175.1 | |||
| XM_011527874.2 | XP_011526176.1 | |||
| XM_011527875.2 | XP_011526177.1 | |||
| XM_011527876.2 | XP_011526178.1 | |||
| XM_011527878.2 | XP_011526180.1 | |||
| XM_011527879.2 | XP_011526181.1 | |||
| BC061700.1 | AAH61700.1 | |||
| DA622182.1 | ||||
| DA628976.1 | ||||
| U27333.1 | AAC50192.1 | |||
| U27335.1 | AAC50194.1 | |||
| U27337.1 | AAC50196.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893160 | Chr.19:5827086 - 5839735 on Build GRCh38 | Loss |
|
| esv3643541 | Chr.19:5831170 - 5844611 on Build GRCh38 | Loss |
|
| nsv1062851 | Chr.19:5531540 - 5893590 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map