Assay Details
Target Gene Details
Entrez Gene ID: | 57402 |
Gene Name: | S100 calcium binding protein A14 |
Gene Aliases: |
BCMP84, S100A15 |
Location: |
Chr.1:153614255-153616917 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100A14 | NM_020672.2 | NP_065723.1 | ||
| XM_005245362.1 | XP_005245419.1 | |||
| XM_017001875.1 | XP_016857364.1 | |||
| AY007220.1 | AAG01893.1 | |||
| BG677973.1 |
More Information
Set Membership: |
|

Genomic Map