Assay Details
Target Gene Details
Entrez Gene ID: | 79139 |
Gene Name: | derlin 1 |
Gene Aliases: |
DER-1, DER1 |
Location: |
Chr.8:123013164-123042423 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DERL1 | NM_001134671.2 | 8 | 2912 | NP_001128143.1 |
| NM_024295.5 | 8 | 2972 | NP_077271.1 | |
| XM_006716657.1 | 7 | 2456 | XP_006716720.1 | |
| XM_011517299.1 | 7 | 2772 | XP_011515601.1 | |
| AA976062.1 | ||||
| AF131854.1 | 1 | 1480 | ||
| AI755206.1 | 1 | 223 | ||
| AY358818.1 | 8 | 2850 | AAQ89177.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1032827 | Chr.8:122655842 - 123258160 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map