Assay Details
Target Gene Details
Entrez Gene ID: | 375612 |
Gene Name: | lipoma HMGIC fusion partner-like 3 |
Gene Aliases: |
LHFPL4 |
Location: |
Chr.7:104328656-104908558 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LHFPL3 | NM_199000.2 | 3 | 1321 | NP_945351.1 |
| XM_005250327.1 | 4 | 1246 | XP_005250384.1 | |
| AK124251.1 | 3 | 1090 | ||
| AK299759.1 | 3 | 1200 | ||
| AY260763.1 | 3 | 1321 | AAP14954.1 | |
| BC129937.1 | 3 | 1277 | ||
| BC129938.1 | 4 | 1325 | ||
| BC129941.1 | 4 | 1325 |
Target Gene Details
Entrez Gene ID: | 723809 |
Gene Name: | LHFPL3 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.7:104894628-104926645 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LHFPL3-AS2 | NR_027374.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv436902 | Chr.7:104802439 - 105205347 on Build GRCh38 | Insertion |
|
| nsv949923 | Chr.7:104904254 - 104908653 on Build GRCh38 | Duplication |
|
| nsv831086 | Chr.7:104782056 - 104956319 on Build GRCh38 | Loss |
|

Genomic Map