Assay Details
Target Gene Details
Entrez Gene ID: | 98 |
Gene Name: | acylphosphatase 2 |
Gene Aliases: |
ACYM, ACYP |
Location: |
Chr.2:53970838-54305300 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACYP2 | NM_001320586.1 | NP_001307515.1 | ||
| NM_001320587.1 | NP_001307516.1 | |||
| NM_001320588.1 | NP_001307517.1 | |||
| NM_001320589.1 | NP_001307518.1 | |||
| NM_138448.3 | NP_612457.1 | |||
| XM_017005411.1 | XP_016860900.1 | |||
| XM_017005412.1 | XP_016860901.1 | |||
| XM_017005413.1 | XP_016860902.1 | |||
| BC012290.1 | AAH12290.1 | |||
| BF671963.1 | ||||
| BG741807.1 | ||||
| BU543943.1 | ||||
| BU857820.1 | ||||
| CA308062.1 | ||||
| X84195.1 | CAA58988.1 |
Target Gene Details
Entrez Gene ID: | 388951 |
Gene Name: | TSPY like 6 |
Gene Aliases: |
- |
Location: |
Chr.2:54253178-54256272 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSPYL6 | NM_001003937.2 | 1 | 722 | NP_001003937.2 |
| AK097417.1 | 1 | 676 | BAC05043.1 | |
| BC068576.1 | 1 | 657 | AAH68576.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011975 | Chr.2:54248729 - 54268085 on Build GRCh38 | Loss |
|
| nsv1004632 | Chr.2:54242146 - 54363343 on Build GRCh38 | Loss |
|
| esv2720078 | Chr.2:53962225 - 55281329 on Build GRCh38 | Deletion |
|
| esv3892403 | Chr.2:54180955 - 54296148 on Build GRCh38 | Loss |
|
| dgv3845n100 | Chr.2:53992912 - 54487416 on Build GRCh38 | Gain |
|
| nsv1009828 | Chr.2:54165985 - 54261584 on Build GRCh38 | Loss |
|
| nsv582012 | Chr.2:54231956 - 54275775 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map