Assay Details
Target Gene Details
Entrez Gene ID: | 983 |
Gene Name: | cyclin dependent kinase 1 |
Gene Aliases: |
CDC2, CDC28A, P34CDC2 |
Location: |
Chr.10:60778331-60794852 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK1 | NM_001320918.1 | NP_001307847.1 | ||
| NM_001786.4 | NP_001777.1 | |||
| NM_033379.4 | NP_203698.1 | |||
| XM_005270303.3 | XP_005270360.1 | |||
| AK291939.1 | ||||
| AW976143.1 | ||||
| BC014563.1 | AAH14563.1 | |||
| BF685285.1 | ||||
| BG723076.1 | ||||
| BT007004.1 | AAP35650.1 | |||
| CR933728.1 | CAI46271.1 | |||
| D88357.1 | BAA26001.1 | |||
| X05360.1 | CAA28963.1 | |||
| Y00272.1 | CAA68376.1 |
More Information
Set Membership: |
|

Genomic Map