Assay Details
Target Gene Details
Entrez Gene ID: | 56300 |
Gene Name: | interleukin 36, gamma |
Gene Aliases: |
IL-1F9, IL-1H1, IL-1RP2, IL1E, IL1F9, IL1H1, IL1RP2 |
Location: |
Chr.2:112978019-112985672 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL36G | NM_001278568.1 | 4 | 850 | NP_001265497.1 |
| NM_019618.3 | 5 | 955 | NP_062564.1 | |
| AF200492.1 | 5 | 950 | AAF69248.1 | |
| AF206696.1 | 5 | 942 | AAG35670.1 |
More Information
Set Membership: |
|

Genomic Map