Assay Details
Target Gene Details
Entrez Gene ID: | 80314 |
Gene Name: | enhancer of polycomb homolog 1 |
Gene Aliases: |
Epl1 |
Location: |
Chr.10:32267716-32378798 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPC1 | NM_001272004.1 | 14 | 2780 | NP_001258933.1 |
| NM_001272019.2 | 14 | 2386 | NP_001258948.1 | |
| NM_001282391.1 | 14 | 2632 | NP_001269320.1 | |
| NM_025209.3 | 15 | 2849 | NP_079485.1 | |
| XM_006717510.1 | 15 | 2701 | XP_006717573.1 | |
| AF277374.1 | 15 | 2816 | AAG41402.1 | |
| AF286905.1 | 14 | 2450 | AAK60501.1 | |
| AK024329.1 | 11 | 1895 | BAB14888.1 | |
| AK092304.1 | 14 | 2386 | BAC03857.1 | |
| AK299676.1 | ||||
| BC036529.1 | 14 | 2781 | AAH36529.1 |
More Information
Set Membership: |
|

Genomic Map