Assay Details
Target Gene Details
Entrez Gene ID: | 5743 |
Gene Name: | prostaglandin-endoperoxide synthase 2 |
Gene Aliases: |
COX-2, COX2, GRIPGHS, PGG/HS, PGHS-2, PHS-2, hCox-2 |
Location: |
Chr.1:186671812-186680427 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGS2 | NM_000963.3 | 10 | 4369 | NP_000954.1 |
| BM978796.1 | 1 | 139 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762854 | Chr.1:186577969 - 186878442 on Build GRCh38 | Gain |
|
| dgv666n54 | Chr.1:186671926 - 186674232 on Build GRCh38 | Loss |
|
| nsv548400 | Chr.1:186671572 - 186673199 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map