Assay Details
Target Gene Details
Entrez Gene ID: | 79816 |
Gene Name: | transducin like enhancer of split 6 |
Gene Aliases: |
GRG6, PREMBL |
Location: |
Chr.19:2977410-2995184 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TLE6 | NM_001143986.1 | NP_001137458.1 | ||
| NM_024760.2 | NP_079036.1 | |||
| XM_005259645.2 | XP_005259702.1 | |||
| XM_011528300.2 | XP_011526602.1 | |||
| AK024071.1 | BAB14815.1 | |||
| BC013986.2 | AAH13986.1 | |||
| BC020206.1 | AAH20206.1 | |||
| DA013757.1 | ||||
| DB059462.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833709 | Chr.19:2950244 - 3068609 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map