Assay Details
Target Gene Details
Entrez Gene ID: | 27329 |
Gene Name: | angiopoietin like 3 |
Gene Aliases: |
ANG-5, ANGPT5, ANL3, FHBL2 |
Location: |
Chr.1:62597487-62606305 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANGPTL3 | NM_014495.3 | 1 | 90 | NP_055310.1 |
| AF152562.1 | AAD34156.1 | |||
| AK289426.1 | 1 | 57 | ||
| AK315304.1 | 1 | 60 | ||
| AY358273.1 | AAQ88640.1 | |||
| BC007059.1 | 1 | 57 | ||
| BC058287.1 | 1 | 61 | AAH58287.1 | |
| BG562798.1 | 1 | 90 | ||
| BG564770.1 | 1 | 62 |
Target Gene Details
Entrez Gene ID: | 85440 |
Gene Name: | dedicator of cytokinesis 7 |
Gene Aliases: |
EIEE23, ZIR2 |
Location: |
Chr.1:62454726-62688368 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOCK7 | NM_001271999.1 | NP_001258928.1 | ||
| NM_001272000.1 | NP_001258929.1 | |||
| NM_001272001.1 | NP_001258930.1 | |||
| NM_001272002.1 | NP_001258931.1 | |||
| NM_033407.3 | NP_212132.2 | |||
| XM_005271292.2 | XP_005271349.1 | |||
| XM_011542326.2 | XP_011540628.1 | |||
| XM_011542327.2 | XP_011540629.1 | |||
| XM_011542328.2 | XP_011540630.1 | |||
| XM_011542330.2 | XP_011540632.1 | |||
| XM_017002639.1 | XP_016858128.1 | |||
| XM_017002640.1 | XP_016858129.1 | |||
| AK055401.1 | BAB70917.1 | |||
| AK292640.1 | ||||
| DQ118679.1 | AAZ38451.1 | |||
| DQ118680.1 | AAZ38452.1 | |||
| DQ309763.1 | ABC33725.1 | |||
| DQ341187.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1000988 | Chr.1:62596120 - 62641855 on Build GRCh38 | Loss |
|
| nsv1014376 | Chr.1:62510801 - 62715688 on Build GRCh38 | Loss |
|
| nsv1011203 | Chr.1:62581375 - 62616208 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map