Assay Details
Target Gene Details
Entrez Gene ID: | 6571 |
Gene Name: | solute carrier family 18 member A2 |
Gene Aliases: |
SVAT, SVMT, VAT2, VMAT2 |
Location: |
Chr.10:117241073-117279430 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC18A2 | NM_003054.4 | 15 | 1557 | NP_003045.2 |
| AK314997.1 | 15 | 1510 | ||
| BC030593.1 | 14 | 1815 | AAH30593.1 | |
| BC108927.1 | 14 | 1422 | AAI08928.1 | |
| BC108928.2 | 14 | 1422 | AAI08929.1 | |
| L09118.1 | 15 | 1499 | AAA59877.1 | |
| L14269.1 | 15 | 1427 | AAA91853.1 | |
| L23205.1 | 15 | 1507 | AAA61290.1 | |
| X71354.1 | 15 | 1457 | CAA50489.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3577760 | Chr.10:117242558 - 117339387 on Build GRCh38 | Gain |
|
| esv3624674 | Chr.10:117141857 - 117425204 on Build GRCh38 | Gain |
|
| nsv1148753 | Chr.10:117256078 - 117271368 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map