Assay Details
Target Gene Details
Entrez Gene ID: | 64184 |
Gene Name: | epididymal protein 3B |
Gene Aliases: |
EP3B, FAM12B, HE3-BETA, HE3B, RAM2 |
Location: |
Chr.14:20768427-20770948 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EDDM3B | NM_022360.4 | 2 | 172 | NP_071755.1 |
| AI075915.1 | ||||
| AY358865.1 | 2 | 172 | AAQ89224.1 | |
| BC128030.1 | 2 | 135 | ||
| BX112699.1 | 2 | 168 | ||
| X76386.2 | 1 | 134 | CAC17141.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759972 | Chr.14:20747949 - 21003455 on Build GRCh38 | Gain+Loss |
|
| esv23438 | Chr.14:20332606 - 20789719 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map