Assay Details
Target Gene Details
Entrez Gene ID: | 4257 |
Gene Name: | microsomal glutathione S-transferase 1 |
Gene Aliases: |
GST12, MGST, MGST-I |
Location: |
Chr.12:16347142-16377189 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGST1 | NM_001260511.1 | NP_001247440.1 | ||
| NM_001260512.1 | NP_001247441.1 | |||
| NM_001267598.1 | NP_001254527.1 | |||
| NM_020300.4 | NP_064696.1 | |||
| NM_145791.2 | NP_665734.1 | |||
| NM_145792.2 | NP_665735.1 | |||
| NR_048545.1 | ||||
| NR_048547.1 | ||||
| XM_006719079.1 | XP_006719142.1 | |||
| XM_011520673.1 | XP_011518975.1 | |||
| XM_011520674.2 | XP_011518976.1 | |||
| AA314967.1 | ||||
| AK291148.1 | ||||
| AK309707.1 | ||||
| AV655242.1 | ||||
| BC005923.1 | AAH05923.1 | |||
| BE568125.1 | ||||
| BG569388.1 | ||||
| BG617746.1 | ||||
| BG778603.1 | ||||
| H49331.1 | ||||
| R86750.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1049805 | Chr.12:16284461 - 16498558 on Build GRCh38 | Loss |
|
| nsv1036638 | Chr.12:15912446 - 16358900 on Build GRCh38 | Loss |
|
| esv3892149 | Chr.12:16266471 - 16354205 on Build GRCh38 | Loss |
|
| esv3892150 | Chr.12:16339581 - 16384735 on Build GRCh38 | Loss |
|
| nsv832338 | Chr.12:16319300 - 16487163 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map