Assay Details
Target Gene Details
Entrez Gene ID: | 54676 |
Gene Name: | GTP binding protein 2 |
Gene Aliases: |
- |
Location: |
Chr.6:43620481-43629246 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTPBP2 | NM_001286216.1 | 12 | 2073 | NP_001273145.1 |
| NM_019096.4 | 12 | 2268 | NP_061969.3 | |
| AB024574.1 | 13 | 2111 | BAB12431.1 | |
| AF168990.1 | 12 | 2065 | AAF78884.1 | |
| AJ420518.1 | 7 | 1431 | ||
| AK000430.1 | BAA91160.1 | |||
| AK290267.1 | ||||
| AL834331.1 | 12 | 2050 | CAD38999.1 | |
| BC020980.2 | 8 | 1684 | AAH20980.2 | |
| BC028347.2 | 10 | 1976 | AAH28347.2 | |
| BC064968.1 | 12 | 2221 | AAH64968.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5281 | Chr.6:43611090 - 43655905 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map