Assay Details
Target Gene Details
Entrez Gene ID: | 64816 |
Gene Name: | cytochrome P450 family 3 subfamily A member 43 |
Gene Aliases: |
- |
Location: |
Chr.7:99828013-99867138 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP3A43 | NM_001278921.1 | 9 | 1124 | NP_001265850.1 |
| NM_022820.4 | 12 | 1457 | NP_073731.1 | |
| NM_057095.2 | 12 | 1454 | NP_476436.1 | |
| NM_057096.3 | NP_476437.1 | |||
| NR_103868.1 | ||||
| NR_103869.1 | 13 | 1575 | ||
| XM_017012542.1 | 10 | 1125 | XP_016868031.1 | |
| XM_017012543.1 | 9 | 1283 | XP_016868032.1 | |
| XM_017012544.1 | 8 | 1011 | XP_016868033.1 | |
| XM_017012545.1 | 8 | 991 | XP_016868034.1 | |
| AA417369.1 | 2 | 266 | ||
| AF280108.1 | 12 | 1351 | AAG33009.1 | |
| AF280109.1 | 12 | 1354 | AAG33010.1 | |
| AF280110.1 | AAG33011.1 | |||
| AF280111.1 | 13 | 1472 | AAG33012.1 | |
| AF280112.1 | 9 | 1031 | AAG33013.1 | |
| AF280113.1 | 9 | 1687 | AAG33014.1 | |
| AF319634.1 | 12 | 1351 | AAK00325.1 | |
| AF337813.1 | 12 | 1454 | AAK38841.1 | |
| AY390423.1 | 12 | 1350 | AAQ92351.1 | |
| AY390424.1 | 12 | 1351 | AAQ92352.1 | |
| AY390425.1 | 12 | 1351 | AAQ92353.1 | |
| AY390426.1 | 12 | 1350 | AAQ92354.1 | |
| BC100981.3 | 9 | 1094 | AAI00982.1 | |
| BC100982.3 | AAI00983.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3614284 | Chr.7:99854025 - 99864973 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map