Assay Details
Target Gene Details
Entrez Gene ID: | 6324 |
Gene Name: | sodium voltage-gated channel beta subunit 1 |
Gene Aliases: |
ATFB13, BRGDA5, GEFSP1 |
Location: |
Chr.19:35030688-35040449 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCN1B | NM_001037.4 | NP_001028.1 | ||
| NM_001321605.1 | NP_001308534.1 | |||
| NM_199037.4 | NP_950238.1 | |||
| AK295477.1 | ||||
| AK313279.1 | ||||
| AY391842.1 | AAR25552.1 | |||
| BC021266.2 | AAH21266.2 | |||
| BC067122.1 | AAH67122.1 | |||
| BM805468.1 | ||||
| BT019923.1 | AAV38726.1 | |||
| DA062026.1 | ||||
| L10338.1 | AAA60391.1 | |||
| L16242.1 | AAA61277.1 |
More Information
Set Membership: |
|

Genomic Map