Assay Details
Target Gene Details
Entrez Gene ID: | 84364 |
Gene Name: | ADP ribosylation factor GTPase activating protein 2 |
Gene Aliases: |
IRZ, NBLA10535, ZFP289, ZNF289 |
Location: |
Chr.11:47164298-47177503 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARFGAP2 | NM_001242832.1 | 15 | 1975 | NP_001229761.1 |
| NM_032389.4 | 16 | 2059 | NP_115765.2 | |
| XM_005253166.1 | 17 | 2554 | XP_005253223.1 | |
| XM_005253167.1 | 17 | 2521 | XP_005253224.1 | |
| XM_005253168.2 | 17 | 2479 | XP_005253225.1 | |
| XM_006718346.1 | 16 | 2512 | XP_006718409.1 | |
| XM_011520405.1 | 16 | 2395 | XP_011518707.1 | |
| XM_017018413.1 | 16 | 2479 | XP_016873902.1 | |
| XM_017018414.1 | 16 | 1875 | XP_016873903.1 | |
| XM_017018415.1 | 16 | 1827 | XP_016873904.1 | |
| AB073358.1 | 15 | 1669 | BAE45716.1 | |
| AK027482.1 | 16 | 1795 | BAB55144.1 | |
| AK098324.1 | 11 | 1405 | ||
| AK129511.1 | 6 | 752 | ||
| AK304541.1 | ||||
| AK315992.1 | ||||
| AL834337.1 | 16 | 1778 | CAD39004.1 | |
| BC030148.2 | 16 | 1802 | AAH30148.1 |
More Information
Set Membership: |
|

Genomic Map