Assay Details
Target Gene Details
Entrez Gene ID: | 85440 |
Gene Name: | dedicator of cytokinesis 7 |
Gene Aliases: |
EIEE23, ZIR2 |
Location: |
Chr.1:62454726-62688368 on Build GRCh38 |
Assay Gene Location: | Within Exon 53 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOCK7 | NM_001271999.1 | 49 | 6658 | NP_001258928.1 |
| NM_001272000.1 | 49 | 6571 | NP_001258929.1 | |
| NM_001272001.1 | 48 | 6565 | NP_001258930.1 | |
| NM_033407.3 | 49 | 6598 | NP_212132.2 | |
| XM_005271292.2 | 50 | 6654 | XP_005271349.1 | |
| XM_011542326.2 | 50 | 6681 | XP_011540628.1 | |
| XM_011542327.2 | 49 | 6675 | XP_011540629.1 | |
| XM_011542328.2 | 49 | 6666 | XP_011540630.1 | |
| XM_017002639.1 | 48 | 6582 | XP_016858128.1 | |
| AB051558.1 | 30 | 4074 | BAB21862.1 | |
| AK055296.1 | 2 | 1867 | ||
| AK055493.1 | 16 | 2215 | BAB70933.1 | |
| AK055905.1 | 21 | 3090 | BAB71042.1 | |
| AK091479.1 | 23 | 3065 | ||
| AK125049.1 | 35 | 4743 | BAC86032.1 | |
| AK299737.1 | 21 | 2775 | ||
| BC016392.1 | 18 | 2562 | ||
| BC112387.1 | 9 | 1317 | ||
| BG190976.1 | 1 | 211 | ||
| BG205481.1 | 1 | 212 | ||
| BG207092.1 | 1 | 216 | ||
| BU567903.1 | ||||
| DQ341187.1 | 49 | 6565 |
More Information
Set Membership: |
|

Genomic Map