Assay Details
Target Gene Details
Entrez Gene ID: | 55361 |
Gene Name: | phosphatidylinositol 4-kinase type 2 alpha |
Gene Aliases: |
PI4KII, PIK42A |
Location: |
Chr.10:97640686-97676434 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PI4K2A | NM_018425.3 | NP_060895.1 | ||
| AJ303098.1 | CAC38065.1 | |||
| BC003167.1 | AAH03167.1 | |||
| BT007330.1 | AAP35994.1 |
More Information
Set Membership: |
|

Genomic Map