Assay Details
Target Gene Details
Entrez Gene ID: | 2494 |
Gene Name: | nuclear receptor subfamily 5 group A member 2 |
Gene Aliases: |
B1F, B1F2, CPF, FTF, FTZ-F1, FTZ-F1beta, LRH-1, LRH1, hB1F-2 |
Location: |
Chr.1:200027602-200177424 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NR5A2 | NM_003822.4 | 1 | 126 | NP_003813.1 |
| NM_205860.2 | 1 | 126 | NP_995582.1 | |
| XM_011509380.1 | XP_011507682.1 | |||
| XM_011509382.1 | XP_011507684.1 | |||
| AB019246.1 | BAA34092.1 | |||
| AF146343.1 | 1 | 86 | AAD37378.1 | |
| BC118571.1 | ||||
| BC118652.1 | ||||
| U93553.1 | 1 | 126 | AAD03155.1 |
More Information
Set Membership: |
|

Genomic Map