Assay Details
Target Gene Details
Entrez Gene ID: | 128360 |
Gene Name: | olfactory receptor family 10 subfamily T member 2 |
Gene Aliases: |
OR1-3 |
Location: |
Chr.1:158398522-158399466 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR10T2 | NM_001004475.1 | 1 | 817 | NP_001004475.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831692 | Chr.1:158253669 - 158422226 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map