Assay Details
Target Gene Details
Entrez Gene ID: | 9420 |
Gene Name: | cytochrome P450 family 7 subfamily B member 1 |
Gene Aliases: |
CBAS3, CP7B, SPG5A |
Location: |
Chr.8:64590851-64798791 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP7B1 | NM_001324112.1 | NP_001311041.1 | ||
| NM_004820.4 | 6 | 1635 | NP_004811.1 | |
| XM_017014002.1 | 7 | 20033 | XP_016869491.1 | |
| AF029403.1 | 6 | 1565 | AAC95426.1 | |
| AF127090.1 | 6 | 1590 | AAD20021.1 | |
| BC029155.1 | ||||
| BC136574.1 | 6 | 1605 | ||
| BM729103.1 | ||||
| BP349825.1 | 2 | 327 |
More Information
Set Membership: |
|

Genomic Map