Assay Details
Target Gene Details
Entrez Gene ID: | 79754 |
Gene Name: | ankyrin repeat and SOCS box containing 13 |
Gene Aliases: |
- |
Location: |
Chr.10:5638857-5666595 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB13 | NM_024701.3 | NP_078977.2 | ||
| NR_024581.1 | ||||
| NR_037164.1 | ||||
| AF403031.1 | AAL57350.1 | |||
| AK023196.1 | BAB14456.1 | |||
| AK091935.1 | ||||
| AK292071.1 | ||||
| BC012056.1 | AAH12056.1 | |||
| BG436652.1 | ||||
| BP249999.1 | ||||
| CR457302.1 | CAG33583.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv19e55 | Chr.10:5623811 - 5674895 on Build GRCh38 | Gain |
|
| nsv442170 | Chr.10:5623499 - 5675164 on Build GRCh38 | Gain |
|
| nsv975791 | Chr.10:5610262 - 5669114 on Build GRCh38 | Duplication |
|
| nsv8599 | Chr.10:5607166 - 5677965 on Build GRCh38 | Gain |
|
| nsv523491 | Chr.10:5500146 - 5678104 on Build GRCh38 | Gain |
|
| nsv470917 | Chr.10:5612204 - 5673438 on Build GRCh38 | Gain |
|
| esv3622232 | Chr.10:5609236 - 5660606 on Build GRCh38 | Gain |
|
| nsv519582 | Chr.10:5604140 - 5678104 on Build GRCh38 | Gain |
|
| esv2760149 | Chr.10:5623495 - 5682419 on Build GRCh38 | Gain |
|
| dgv665n100 | Chr.10:5607797 - 5683245 on Build GRCh38 | Gain |
|
| nsv549892 | Chr.10:5610314 - 5673438 on Build GRCh38 | Gain |
|
| esv2759729 | Chr.10:5515061 - 5762817 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map