Assay Details
Target Gene Details
Entrez Gene ID: | 64343 |
Gene Name: | 5-azacytidine induced 2 |
Gene Aliases: |
AZ2, NAP1, TILP |
Location: |
Chr.3:28322353-28349129 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AZI2 | NM_001134432.1 | NP_001127904.1 | ||
| NM_001134433.1 | NP_001127905.1 | |||
| NM_001271650.1 | NP_001258579.1 | |||
| NM_022461.4 | NP_071906.1 | |||
| XM_005265392.2 | XP_005265449.1 | |||
| XM_011534009.2 | XP_011532311.1 | |||
| XM_011534010.2 | XP_011532312.1 | |||
| XM_011534011.2 | XP_011532313.1 | |||
| AK025592.1 | BAB15182.1 | |||
| AK290637.1 | ||||
| AY151386.1 | AAO05967.1 | |||
| BM546093.1 | ||||
| BX327318.2 | ||||
| BX648471.1 | ||||
| CB995808.1 | ||||
| CF528511.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3568668 | Chr.3:28319282 - 28351354 on Build GRCh38 | Loss |
|
| dgv1365e212 | Chr.3:28331663 - 28360829 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map