Assay Details
Target Gene Details
Entrez Gene ID: | 80124 |
Gene Name: | valosin containing protein interacting protein 1 |
Gene Aliases: |
DUBA3, VCIP135 |
Location: |
Chr.8:66630253-66667217 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VCPIP1 | NM_025054.4 | 3 | 6550 | NP_079330.2 |
| AB058753.1 | 3 | 6550 | ||
| AK297345.1 | ||||
| BC049379.1 | 3 | 4980 | AAH49379.1 | |
| CR936659.1 | 3 | 6581 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831346 | Chr.8:66574320 - 66763089 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map