Assay Details
Target Gene Details
Entrez Gene ID: | 10635 |
Gene Name: | RAD51 associated protein 1 |
Gene Aliases: |
PIR51 |
Location: |
Chr.12:4538784-4560047 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD51AP1 | NM_001130862.1 | 10 | 1441 | NP_001124334.1 |
| NM_006479.4 | 9 | 1390 | NP_006470.1 | |
| AF006259.1 | AAC39554.1 | |||
| AK096930.1 | 10 | 1318 | BAC04902.1 | |
| AK290428.1 | 10 | 1335 | ||
| AK291948.1 | 9 | 1284 | ||
| AK300773.1 | 8 | 1222 | ||
| AK309706.1 | 3 | 561 | ||
| AL832243.1 | 10 | 1319 | ||
| BC006992.1 | 9 | 1284 | AAH06992.1 | |
| BC016330.1 | 10 | 1333 | AAH16330.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483060 | Chr.12:3190835 - 5290834 on Build GRCh38 | Loss |
|
| nsv832318 | Chr.12:4546633 - 4766671 on Build GRCh38 | Gain |
|
| nsv557123 | Chr.12:4327466 - 4627403 on Build GRCh38 | Gain |
|
| nsv1040098 | Chr.12:3987637 - 5646161 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map