Assay Details
Target Gene Details
Entrez Gene ID: | 85440 |
Gene Name: | dedicator of cytokinesis 7 |
Gene Aliases: |
EIEE23, ZIR2 |
Location: |
Chr.1:62454726-62688368 on Build GRCh38 |
Assay Gene Location: | Within Exon 42 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOCK7 | NM_001271999.1 | 41 | 5373 | NP_001258928.1 |
| NM_001272000.1 | 40 | 5280 | NP_001258929.1 | |
| NM_001272001.1 | 40 | 5280 | NP_001258930.1 | |
| NM_033407.3 | 40 | 5307 | NP_212132.2 | |
| XM_005271292.2 | 41 | 5363 | XP_005271349.1 | |
| XM_011542326.2 | 41 | 5390 | XP_011540628.1 | |
| XM_011542327.2 | 41 | 5390 | XP_011540629.1 | |
| XM_011542328.2 | 41 | 5390 | XP_011540630.1 | |
| XM_011542330.2 | 41 | 5390 | XP_011540632.1 | |
| XM_017002639.1 | 40 | 5297 | XP_016858128.1 | |
| XM_017002640.1 | 41 | 5390 | XP_016858129.1 | |
| AB051558.1 | 21 | 2783 | BAB21862.1 | |
| AK022097.1 | 2 | 1103 | ||
| AK055493.1 | 7 | 924 | BAB70933.1 | |
| AK055905.1 | 13 | 1806 | BAB71042.1 | |
| AK091479.1 | 14 | 1774 | ||
| AK125049.1 | 26 | 3452 | BAC86032.1 | |
| AK299737.1 | 12 | 1490 | ||
| BC016392.1 | 10 | 1277 | ||
| BC112387.1 | ||||
| DQ118679.1 | 37 | 4852 | AAZ38451.1 | |
| DQ118680.1 | 37 | 4852 | AAZ38452.1 | |
| DQ341187.1 | 41 | 5280 |
More Information
Set Membership: |
|

Genomic Map