Assay Details
Target Gene Details
Entrez Gene ID: | 3061 |
Gene Name: | hypocretin receptor 1 |
Gene Aliases: |
OX1R |
Location: |
Chr.1:31617700-31632518 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HCRTR1 | XM_017001107.1 | XP_016856596.1 |
Target Gene Details
Entrez Gene ID: | 553115 |
Gene Name: | penta-EF-hand domain containing 1 |
Gene Aliases: |
ABP32, PEF1A |
Location: |
Chr.1:31629862-31645237 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PEF1 | NM_012392.3 | 5 | 1707 | NP_036524.1 |
| NR_033686.1 | 4 | 1406 | ||
| NR_033688.1 | 5 | 1548 | ||
| XM_011541745.1 | 5 | 1334 | XP_011540047.1 | |
| XM_011541746.2 | 5 | 1801 | XP_011540048.1 | |
| XM_011541747.1 | 5 | 1676 | XP_011540049.1 | |
| XM_017001680.1 | 5 | 1665 | XP_016857169.1 | |
| XM_017001681.1 | 5 | 2972 | XP_016857170.1 | |
| AB018357.1 | 5 | 1346 | BAA84922.1 | |
| AB026628.1 | 5 | 1346 | BAA85163.1 | |
| AK001420.1 | 5 | 1359 | BAA91680.1 | |
| AK092483.1 | 1 | 2701 | ||
| AY359011.1 | 5 | 1350 | AAQ89370.1 | |
| BC002773.2 | 5 | 1336 | AAH02773.1 | |
| BC012561.1 | 5 | 1352 | AAH12561.1 | |
| CA307778.1 | 1 | 291 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545951 | Chr.1:31626924 - 31699894 on Build GRCh38 | Gain |
|
| nsv1000976 | Chr.1:31612766 - 31700055 on Build GRCh38 | Loss |
|
| esv2758930 | Chr.1:31209533 - 32000730 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map