Assay Details
Target Gene Details
Entrez Gene ID: | 6717 |
Gene Name: | sorcin |
Gene Aliases: |
CP-22, CP22, SCN, V19 |
Location: |
Chr.7:88205115-88226993 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRI | NM_003130.3 | 8 | 679 | NP_003121.1 |
| NM_198901.1 | 8 | 655 | NP_944490.1 | |
| XM_011516528.1 | 7 | 549 | XP_011514830.1 | |
| AK129770.1 | 8 | 648 | ||
| AK295225.1 | 8 | 512 | ||
| BC011025.1 | 8 | 637 | AAH11025.1 | |
| BI559592.1 | 8 | 673 | ||
| BI597920.1 | 8 | 675 | ||
| BM739144.1 | ||||
| BM793972.1 | 5 | 397 | ||
| BU854353.1 | 4 | 318 | ||
| L12387.1 | 8 | 625 | AAA92155.1 | |
| M32886.1 | 8 | 633 | AAA60588.1 |
More Information
Set Membership: |
|

Genomic Map