Assay Details
Target Gene Details
Entrez Gene ID: | 79637 |
Gene Name: | armadillo repeat containing 7 |
Gene Aliases: |
- |
Location: |
Chr.17:75109952-75130265 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARMC7 | NM_001304271.1 | NP_001291200.1 | ||
| NM_024585.3 | NP_078861.1 | |||
| AK025813.1 | BAB15242.1 | |||
| BC011728.1 | AAH11728.1 |
Target Gene Details
Entrez Gene ID: | 30833 |
Gene Name: | 5', 3'-nucleotidase, cytosolic |
Gene Aliases: |
DNT, DNT1, HEL74, P5N2, PN-I, PN-II, UMPH2, cdN, dNT-1 |
Location: |
Chr.17:75130225-75131965 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NT5C | NM_001252377.1 | NP_001239306.1 | ||
| NM_014595.2 | NP_055410.1 | |||
| NR_045513.1 | ||||
| AF154829.2 | AAF36534.2 | |||
| AK000419.1 | BAA91151.1 | |||
| BC008183.1 | AAH08183.1 | |||
| BC017454.1 | AAH17454.1 | |||
| BC022334.1 | AAH22334.1 | |||
| BG034032.1 | ||||
| BU627693.1 | ||||
| EU794640.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470602 | Chr.17:75090871 - 75208042 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map