Assay Details
Target Gene Details
Entrez Gene ID: | 55192 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C17 |
Gene Aliases: |
- |
Location: |
Chr.15:40767448-40807478 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC17 | NM_018163.2 | NP_060633.1 | ||
| AK001496.1 | BAA91724.1 | |||
| AK308659.1 | ||||
| AK308858.1 | ||||
| AK309307.1 | ||||
| AK310140.1 | ||||
| BC000048.2 | AAH00048.1 | |||
| DA341962.1 |
Target Gene Details
Entrez Gene ID: | 84936 |
Gene Name: | zinc finger FYVE-type containing 19 |
Gene Aliases: |
ANCHR, MPFYVE |
Location: |
Chr.15:40807076-40814569 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFYVE19 | NM_001077268.1 | 1 | 407 | NP_001070736.1 |
| NM_001258420.1 | 1 | 407 | NP_001245349.1 | |
| NM_001258421.1 | 1 | 407 | NP_001245350.1 | |
| NM_032850.4 | NP_116239.3 | |||
| XM_017022684.1 | 1 | 394 | XP_016878173.1 | |
| AF445414.1 | AAO73862.1 | |||
| AK122779.1 | 1 | 394 | ||
| DA611931.1 | 1 | 397 | ||
| DA653225.1 | 1 | 407 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054764 | Chr.15:40776427 - 40904212 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map