Assay Details
Target Gene Details
Entrez Gene ID: | 6853 |
Gene Name: | synapsin I |
Gene Aliases: |
SYN1a, SYN1b, SYNI |
Location: |
Chr.X:47571901-47619857 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYN1 | NM_006950.3 | NP_008881.2 | ||
| NM_133499.2 | NP_598006.1 |
Target Gene Details
Entrez Gene ID: | 7076 |
Gene Name: | TIMP metallopeptidase inhibitor 1 |
Gene Aliases: |
CLGI, EPA, EPO, HCI, TIMP, TIMP-1 |
Location: |
Chr.X:47582291-47586791 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TIMP1 | NM_003254.2 | 1 | 93 | NP_003245.1 |
| XM_017029766.1 | 1 | 47 | XP_016885255.1 | |
| AK074854.1 | 1 | 71 | ||
| AK296116.1 | 1 | 47 | ||
| BC000866.1 | 1 | 46 | AAH00866.1 |
More Information
Set Membership: |
|

Genomic Map