Assay Details
Target Gene Details
Entrez Gene ID: | 341152 |
Gene Name: | olfactory receptor family 2 subfamily AT member 4 |
Gene Aliases: |
OR11-265 |
Location: |
Chr.11:75088692-75100854 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR2AT4 | NM_001005285.1 | 1 | 694 | NP_001005285.1 |
| XM_017017655.1 | 2 | 2334 | XP_016873144.1 | |
| XM_017017656.1 | 2 | 2330 | XP_016873145.1 | |
| XM_017017657.1 | 2 | 1872 | XP_016873146.1 | |
| XM_017017658.1 | 2 | 2357 | XP_016873147.1 | |
| BC136958.1 | 1 | 735 | ||
| BC136959.1 | 1 | 735 |
More Information
Additional Information:
For this assay, SNP(s) [rs111801887] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map