Assay Details
Target Gene Details
Entrez Gene ID: | 8509 |
Gene Name: | N-deacetylase and N-sulfotransferase 2 |
Gene Aliases: |
HSST2, NST2 |
Location: |
Chr.10:73801911-73811831 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDST2 | NM_003635.3 | 7 | 2332 | NP_003626.1 |
| XM_005270255.3 | 7 | 2332 | XP_005270312.1 | |
| XM_005270256.3 | 4 | 537 | XP_005270313.1 | |
| XM_011540310.2 | 6 | 2901 | XP_011538612.1 | |
| XM_017016857.1 | 7 | 2361 | XP_016872346.1 | |
| AB208870.1 | 3 | 1967 | BAD92107.1 | |
| AK293527.1 | 3 | 389 | ||
| AK298957.1 | 5 | 625 | ||
| AK300520.1 | 3 | 535 | ||
| AK301251.1 | ||||
| AK303648.1 | 8 | 2130 | ||
| AK310580.1 | 3 | 498 | ||
| BC035711.1 | 7 | 2085 | AAH35711.1 | |
| BC110588.1 | 5 | 1564 | AAI10589.1 | |
| BC110589.1 | 5 | 1564 | AAI10590.1 | |
| U36601.1 | 5 | 1555 | AAC27120.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv46n21 | Chr.10:73790158 - 73933937 on Build GRCh38 | Loss |
|
| nsv551517 | Chr.10:73799109 - 73898823 on Build GRCh38 | Loss |
|
| nsv831912 | Chr.10:73649351 - 73840445 on Build GRCh38 | Gain |
|
| nsv1035634 | Chr.10:73724401 - 73907300 on Build GRCh38 | Gain |
|
| nsv825468 | Chr.10:73715689 - 73922563 on Build GRCh38 | Gain |
|
| nsv551516 | Chr.10:73749515 - 73847410 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map