Assay Details
Target Gene Details
Entrez Gene ID: | 84186 |
Gene Name: | zinc finger CCHC-type containing 7 |
Gene Aliases: |
AIR1, HSPC086 |
Location: |
Chr.9:37120169-37358149 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZCCHC7 | NM_001289119.1 | 2 | 621 | NP_001276048.1 |
| NM_001289120.1 | 2 | 644 | NP_001276049.1 | |
| NM_001289121.1 | 2 | 762 | NP_001276050.1 | |
| NM_032226.2 | 2 | 617 | NP_115602.2 | |
| NR_110317.1 | ||||
| XM_005251608.4 | 2 | 1526 | XP_005251665.1 | |
| XM_005251612.2 | 2 | 584 | XP_005251669.1 | |
| XM_011518050.1 | 2 | 612 | XP_011516352.1 | |
| XM_017015197.1 | 2 | 739 | XP_016870686.1 | |
| XM_017015198.1 | 2 | 1225 | XP_016870687.1 | |
| XM_017015199.1 | 2 | 586 | XP_016870688.1 | |
| XM_017015200.1 | 2 | 859 | XP_016870689.1 | |
| XM_017015201.1 | XP_016870690.1 | |||
| AK026264.1 | 2 | 507 | BAB15418.1 | |
| AK074608.1 | 1 | 106 | BAC11087.1 | |
| AK303193.1 | ||||
| AK315128.1 | 2 | 633 | ||
| BC005990.1 | 2 | 550 | ||
| BC022434.1 | 3 | 710 | AAH22434.1 | |
| BC036940.1 | 2 | 735 | AAH36940.1 | |
| BP216150.1 | ||||
| BP246540.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv529027 | Chr.9:37108500 - 37353255 on Build GRCh38 | Loss |
|
| nsv466390 | Chr.9:37058138 - 37289935 on Build GRCh38 | Loss |
|
| nsv520965 | Chr.9:37115199 - 37154796 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map