Assay Details
Target Gene Details
Entrez Gene ID: | 55324 |
Gene Name: | ATP binding cassette subfamily F member 3 |
Gene Aliases: |
EST201864 |
Location: |
Chr.3:184186075-184194012 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCF3 | NM_018358.2 | NP_060828.2 | ||
| XM_011512959.2 | XP_011511261.1 | |||
| AK002060.1 | BAA92063.1 | |||
| AK027288.1 | ||||
| AK092415.1 | BAC03881.1 | |||
| AK097073.1 | ||||
| AK290106.1 | ||||
| AK299256.1 | ||||
| AK299433.1 | ||||
| BC009253.2 | AAH09253.1 | |||
| BC051754.2 | AAH51754.1 | |||
| BC051884.1 | ||||
| DB079392.1 | ||||
| DC318082.1 | ||||
| KU178694.1 | ||||
| KU178695.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829812 | Chr.3:184136655 - 184302302 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map