Assay Details
Target Gene Details
Entrez Gene ID: | 1136 |
Gene Name: | cholinergic receptor nicotinic alpha 3 subunit |
Gene Aliases: |
LNCR2, NACHRA3, PAOD2 |
Location: |
Chr.15:78593052-78621295 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRNA3 | NM_000743.4 | 6 | 2488 | NP_000734.2 |
| NM_001166694.1 | NP_001160166.1 | |||
| NR_046313.1 | ||||
| XM_006720382.2 | 6 | 2430 | XP_006720445.1 | |
| AJ584707.1 | CAE48369.1 | |||
| AJ584708.1 | CAE48370.1 | |||
| AJ584709.1 | 1 | 598 | CAE48371.1 | |
| BC000513.2 | 6 | 2173 | AAH00513.1 | |
| BC001642.2 | 6 | 2173 | AAH01642.1 | |
| BC002996.1 | 6 | 2173 | AAH02996.1 | |
| BC006114.1 | AAH06114.1 | |||
| BC098443.1 | 6 | 2369 | AAH98443.1 | |
| BT006646.1 | AAP35292.1 | |||
| M37981.1 | AAA59942.1 |
More Information
Set Membership: |
|

Genomic Map