Assay Details
Target Gene Details
Entrez Gene ID: | 9420 |
Gene Name: | cytochrome P450 family 7 subfamily B member 1 |
Gene Aliases: |
CBAS3, CP7B, SPG5A |
Location: |
Chr.8:64590851-64798791 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP7B1 | NM_001324112.1 | 3 | 733 | NP_001311041.1 |
| NM_004820.4 | 3 | 733 | NP_004811.1 | |
| XM_017014002.1 | 4 | 19131 | XP_016869491.1 | |
| AF029403.1 | 3 | 663 | AAC95426.1 | |
| AF127090.1 | 3 | 688 | AAD20021.1 | |
| BC029155.1 | 3 | 660 | ||
| BC136574.1 | 3 | 703 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv611449 | Chr.8:64598475 - 64630844 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map